Spatel, vibrasjonsspatel
Leverandør: BOCHEM
3540
231-1289EA 620 NOK
231-1289
Spatel, vibrasjonsspatel
Spatler
Stainless steel, plastic shaft. A toothed wheel is turned by a finger making the spatula blade vibrate. This action allows for accurate adding of material.