Tongue depressor
Leverandør: DELTALAB
441142.1 441142.2
231-2645EA 705 NOK
231-2645 231-2646
Tongue depressor
Tungespatel
PS, cream. Tongue depressor with smooth rounded edges.
  • Odour- and taste-free
  • Maintains a high tensile strength, more flexible than wood depressors
  • Features a ribbed finger rest for easy handling
Can also be used for spreading balms and ointments, for stirring liquids or mixing medicine.
NB: *Produktet/produktene på denne siden er utenfor omfanget av EU -direktivet 2019/904