Kobber(II)sulfat pentahydrat ≥98.5%, renset

Leverandør: VWR Chemicals
Warning

23172.293EA 1200 NOK
23172.293 23172.362
Kobber(II)sulfat pentahydrat ≥98.5%, renset
Kobber(II)sulfat pentahydrat
Formel: CuSO₄·5H₂O
Tetthet: 2,284 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149681
CAS-nummer: 7758-99-8
EINECS: 231-847-6
FN: 3077
ADR: 9,III
REACH: 01-2119520566-40
Merck Indeks: 13,02682

Bestill nå

Spesifikasjon Testresultater

Assay Min. 98.5 %
Cl (Chloride) Max. 20 ppm
Fe (Iron) Max. 100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR