Kobber(II)sulfat pentahydrat 99.0-102.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytisk reagens

Leverandør: VWR Chemicals
Danger

23174.233EA 640 NOK
23174.233 23174.290 23174.360 23174.460
Kobber(II)sulfat pentahydrat 99.0-102.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytisk reagens
Kobber(II)sulfat pentahydrat
Formel: CuSO₄·5H₂O
Tetthet: 2,284 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149681
CAS-nummer: 7758-99-8
EINECS: 231-847-6
FN: 3077
ADR: 9,III
REACH: 01-2119520566-40
Merck Indeks: 13,02682

Bestill nå

Spesifikasjon Testresultater

Assay 99.0 - 102.0 %
Insolubility in water Max. 50 ppm
Total N (Nitrogen) Max. 10 ppm
Cl (Chloride) Max. 5 ppm
Ca (Calcium) Max. 50 ppm
Fe (Iron) Max. 20 ppm
K (Potassium) Max. 10 ppm
Mg (Magnesium) Max. 50 ppm
Na (Sodium) Max. 50 ppm
Ni (Nickel) Max. 50 ppm
Pb (Lead) Max. 50 ppm
Zn (Zinc) Max. 0.03 %
Conforms to ACS Passes test
Conforms to Reag. Ph.Eur. Passes test

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR