Bone cutters
Leverandør: MURRAY S
D631/01 D632/01
233-0010EA 1425 NOK
233-0010 233-0160
Bone cutters
Bone Cutters and Rongeurs
Carbon steel, nickel plated. With overlapping spring or coil spring handle.
  • Excellent thermal resistance
  • Easy to clean
  • Safe, simple operation