Clioquinol, Calbiochem®

Leverandør: Merck Millipore Calbiochem
Danger

Synonymer: 5-Chloro-7-iodo-8-quinolinol, 5-Chloro-8-hydroxy-7-iodoquinoline, Clioquinol

233165-1
233165-1EA 1325 NOK
233165-1
Clioquinol, Calbiochem®
Clioquinol
Formel: C₉H₅ClINO
MW: 305,5 g/mol
Kokepunkt: 365 °C (1013 hPa)
Smeltepunkt: 183…184 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00006787
CAS-nummer: 130-26-7
EINECS: 204-984-4
FN: 2811
ADR: 6.1,III
Merck Indeks: 12,05052

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR