2,6-Dichlorophenolindophenol sodium salt hydrate ACS analytical reagent

Leverandør: VWR Chemicals

Synonymer: DPIP, DCIP

23347.105EA 1775 NOK
23347.105
2,6-Dichlorophenolindophenol sodium salt hydrate ACS analytical reagent
2,6-Dichlorophenolindophenol sodium salt hydrate
Formel: C₁₂H₆Cl₂NNaO₂·xH₂O
MW: 290.08 g/mol (anhy)
Smeltepunkt: > 300 °C
MDL nummer: MFCD00150014
CAS-nummer: 1266615-56-8
EINECS: 210-640-4

Bestill nå

Spesifikasjon Testresultater

IR Spectrum Passes test
Interfering dyestuffs Passes test ACS
Loss on drying (120°C) Max. 12 %
Conforms to ACS Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR