Diklormetan, Wasser_0,01 99 Stabilised, GPR RECTAPUR® zS

Leverandør: VWR Chemicals
Warning

Synonymer: Methylene chloride

Stabilisering: 0.1 % EtOH

23349.363EA 1575 NOK
23349.363
Diklormetan, Wasser_0,01 99 Stabilised, GPR RECTAPUR® zS
Diklormetan
Formel: CH₂Cl₂
MW: 84.93 g/mol
Kokepunkt: 39.8 °C (1013 hPa)
Smeltepunkt: –95 °C
Tetthet: 1.322 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00000881
CAS-nummer: 75-09-2
EINECS: 200-838-9
FN: 1593
ADR: 6.1 III
REACH: 01-2119480404-41
Merck Indeks: 12,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay Min. 99 %
Evaporation residue Max. 50 ppm
Water Max. 100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR