Quinoline yellow 95% (dye content), Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY
Warning

Synonymer: Quinoline yellow 2SF, Solvent yellow 33, 2-(2-Quinolinyl)-1H-indene-1,3(2H)-dione

234133-25G
234133-25GEA 619 NOK
234133-25G
Quinoline yellow 95% (dye content), Sigma-Aldrich®
Quinoline yellow

Quinoline Yellow, Dye content 95 % (Solvent yellow 33)

Formel: C₁₈H₁₁NO₂
MW: 273.29 g/mol
Smeltepunkt: 240 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011593
CAS-nummer: 8003-22-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7039556c5469a436b9eadcef53f8703fea813d20&vid=6fcb6e9837f120792f1d297f7538bbd4e8d829c5&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR