Dimetylglyoksim ≥99.0%, AnalaR NORMAPUR® analytical reagent

Leverandør: VWR Chemicals
Danger

Synonymer: Diacetyl glyoxime, Diacetyl dioxime, 2,3-Butanedione dioxime

23474.150EA 1550 NOK
23474.150
Dimetylglyoksim ≥99.0%, AnalaR NORMAPUR® analytical reagent
Dimetylglyoksim
Formel: CH₃C(=NOH)C(=NOH)CH₃
MW: 116.12 g/mol
Kokepunkt: 53 °C (8 torr)
Smeltepunkt: 240…241 °C
Tetthet: 1.37 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002117
CAS-nummer: 95-45-4
EINECS: 202-420-1
Merck Indeks: 13,03272

Bestill nå

Spesifikasjon Testresultater

Assay Min. 99.0 %
Melting point 238 - 242 °C
Ignition residue (SO4) Max. 0.05 %
Insolubility in ethanol 96 % vol Max. 0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR