Dimetylsulfoksid, Wasser_0,01 99,5, AnalaR NORMAPUR® zA

Leverandør: VWR Chemicals

Synonymer: Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO

23488.237EA 0 NOK
23488.237 23488.294
Dimetylsulfoksid, Wasser_0,01 99,5, AnalaR NORMAPUR® zA
Dimetylsulfoksid
Formel: (CH₃)₂SO
MW: 78.14 g/mol
Kokepunkt: 189
Smeltepunkt: 18.5 °C
Tetthet: 1.101 g/cm³ (20 °C)
Flash punkt: 87
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002089
CAS-nummer: 67-68-5
EINECS: 200-664-3
REACH: 01-2119431362-50
Merck Indeks: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay (calculated on anhydrous) Min. 99.5 %
Fe (Iron) Max. 0.0001 %
Heavy metals (as Pb) Max. 0.00010 %
Residue on evaporation Max. 0.0010 %
Water Max. 0.0100 %

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR