1,5-Difenylkarbazid ≥97.0% analytisk reagens

Leverandør: VWR Chemicals
Warning

23550.120EA 460 NOK
23550.120 23550.153
1,5-Difenylkarbazid ≥97.0% analytisk reagens
1,5-Difenylkarbazid
Formel: C₁₃H₁₄N₄O
Kokepunkt: 454 °C (1013 hPa)
Smeltepunkt: 170…172 °C
Tetthet: 1,31 g/cm³ (20 °C)
MDL nummer: MFCD00003013
CAS-nummer: 140-22-7
EINECS: 205-403-7
Merck Indeks: 13,03353
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=03cf8836a36f1077443ec2e3cfa151621925bd46&vid=a1b2bd876c3fbd9d6ded95c8430e4ec16bf5172b&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay Min. 97.0 %
Suited for Cr6+ reagent Passes test
Melting point 171 - 173 °C
Ignition residue (SO4) Max. 0.1 %
Insolubility in methanol Max. 100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR