Ditizon 98,0, AnalaR NORMAPUR® zA

Leverandør: VWR Chemicals
Warning

Synonymer: Dithizone,for analysis, 1,5-Diphenylthiocarbazone

23570.106EA 680 NOK
23570.106
Ditizon 98,0, AnalaR NORMAPUR® zA
Ditizon
Formel: C₆H₅NHNHCSN=NC₆H₅
MW: 256.33 g/mol
Kokepunkt: 406
Smeltepunkt: 168
MDL nummer: MFCD00003025
CAS-nummer: 60-10-6
EINECS: 200-454-1
Merck Indeks: 13,03413

Bestill nå

Spesifikasjon Testresultater

Assay ≥ 98.0 %
Appearance Purple to dark purple or black powder
Specific absorptivity 1% / 1cm (at 605 nm) min. 1522
Relative absorption (A 605nm/at 445nm; Chloroform) min. 2.5
Sulphated ash max. 0.2%
Heavy metals (as Pb) max. 0.001%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR