Silikagel, Pore size: 60 Å ≥99%, powder 200 - 425 mesh, high purity (Davisil Grade 633), Supelco®

Leverandør: Merck


M-Clarity™ quality level = MQ100

236772-100G 236772-1KG
SIAL236772-100GEA 1190 NOK
SIAL236772-100G 236772-1KG
Silikagel, Pore size: 60 Å ≥99%, powder 200 - 425 mesh, high purity (Davisil Grade 633), Supelco®
Silikagel

Silica gel is a rigid 3D network of colloidal silica.


It is exclusively used as a support material for the active titanium(III) centers in ziegler-natta catalysts. Silica gel allows a controlled fragmentation resulting in the formation of uniform polymer particles with narrow particle size distribution and high bulk density. Silica gel was used to compare MCM-41 type mesoporous adsorbents.

Formel: SiO₂
Smeltepunkt: >1600 °C
MDL nummer: MFCD00011232
CAS-nummer: 63231-67-4
EINECS: 231-545-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e2561c187b532735911309fdea49234aeac734f2&vid=356f010c16846a15d1c427b5ff1341a3172bb4b4&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance (Colour) White
Appearance (Form) Powder
Particle Size: 200 - 425 Mesh Conforms
Assay ≥99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR