Tinn(II)klorid dihydrat ≥95%, TECHNICAL

Leverandør: VWR Chemicals
Danger

23738.232EA 600 NOK
23738.232
Tinn(II)klorid dihydrat ≥95%, TECHNICAL
Tinn(II)klorid dihydrat
Formel: SnCl₂·2H₂O
Kokepunkt: 623 °C (1013 hPa)
Smeltepunkt: 246 °C
Tetthet: 2,71 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149863
CAS-nummer: 10025-69-1
EINECS: 231-868-0
FN: 3260
ADR: 8,III
Merck Indeks: 13,08861

Bestill nå

Spesifikasjon Testresultater

Assay Min. 95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR