Etanolamin ≥99%, GPR RECTAPUR® for liquid scintillation counting

Leverandør: VWR Chemicals
Danger

Synonymer: 2-Aminoethanol, Monoethanolamine

23782.291EA 645 NOK
23782.291
Etanolamin ≥99%, GPR RECTAPUR® for liquid scintillation counting
Etanolamin
Formel: NH₂CH₂CH₂OH
MW: 61.08 g/mol
Kokepunkt: 171
Smeltepunkt: 10.5 °C
Tetthet: 1.0164 g/cm³ (20 °C)
Flash punkt: 93
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008183
CAS-nummer: 141-43-5
EINECS: 205-483-3
FN: 2491
ADR: 8 III
Merck Indeks: 13,03762
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82bfb6ea7cf5dde341c60ff9cf752bc5c422eb28&vid=b387e8c39029da4ea050ba6dcfb6123d81ab03c8&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay Min. 99 %
Identification Passes test
Boiling point 170 - 172 °C
Density (20/4) 1.010 - 1.020
Solidification point 9.0 - 10.5 °C
Water Max. 1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR