Kadmium(II)kllorid hemipentahydrat 79.5-81.0% ACS, Fluka™

Leverandør: Honeywell Chemicals
Danger

239208-500G
239208-500GEA 5050 NOK
239208-500G
Kadmium(II)kllorid hemipentahydrat 79.5-81.0% ACS, Fluka™
Kadmium(II)kllorid hemipentahydrat
Formel: CdCl₂·2.5H₂O
MW: 228.35 g/mol
Smeltepunkt: 568
Tetthet: 3.327 g/cm³
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00799071
CAS-nummer: 7790-78-5
EINECS: 233-296-7
FN: 2570
ADR: 6.1 III
Merck Indeks: 13,01617

Bestill nå

Spesifikasjon Testresultater

Vapor Pressure 10 mmHg ( 656 °C)
Assay of CdCl₂ 79.5 - 81.0 %
Water insoluble matter Max. 0.005 %
Calcium (Ca) Max. 0.005 %
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 5 ppm
Potassium (K) Max. 0.02 %
Sodium (Na) Max. 0.05 %
Lead (Pb) Max. 0.005 %
Zinc (Zn) Max. 0.05 %
Ammonium (NH₄) Max. 0.005 %
Nitrate, nitrite (as NO₃) Max. 0.003 %
Sulfate (SO₄) Max. 0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR