Kobalt(II)nitrat heksahydrat ≥98% ACS, Fluka™

Leverandør: Honeywell Chemicals
Danger

239267-100G 239267-500G
239267-100GEA 1375 NOK
239267-100G 239267-500G
Kobalt(II)nitrat heksahydrat ≥98% ACS, Fluka™
Kobalt(II)nitrat heksahydrat
Formel: Co(NO₃)₂·6H₂O
Kokepunkt: Decomposes on heating
Smeltepunkt: 55-56 °C
Tetthet: 1,87 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149647
CAS-nummer: 10026-22-9
EINECS: 233-402-1
FN: 1477
ADR: 5.1,II
Merck Indeks: 13,02469

Bestill nå

Spesifikasjon Testresultater

Appearance of the substance complying
Assay 98.0 - 102.0 %
Water insoluble matter Max. 0.01 %
Calcium (Ca) Max. 0.005 %
Copper (Cu) Max. 0.002 %
Iron (Fe) Max. 0.001 %
Potassium (K) Max. 0.01 %
Magnesium (Mg) Max. 0.005 %
Sodium (Na) Max. 0.05 %
Nickel (Ni) Max. 0.15 %
Lead (Pb) Max. 0.002 %
Zinc (Zn) Max. 0.01 %
Chloride (Cl) Max. 0.002 %
Sulfate (SO₄) Max. 0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR