Natriumsulfat, anhydrous ≥99.0%, granular ACS, Fluka™

Leverandør: Honeywell Chemicals

Synonymer: Disodium sulphate, Bisodium sulphate, Sulphuric acid disodium salt

239313-1KG 239313-2.5KG 239313-500G 239313-5KG 239313-12KG
239313-1KGEA 575 NOK
239313-1KG 239313-2.5KG 239313-500G 239313-5KG 239313-12KG
Natriumsulfat, anhydrous ≥99.0%, granular ACS, Fluka™
Sodium sulphate
Formel: Na₂SO₄
MW: 142.04 g/mol
Kokepunkt: 1429
Smeltepunkt: 884 °C
Tetthet: 2.68 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003504
CAS-nummer: 7757-82-6
EINECS: 231-820-9
Merck Indeks: 12,08829
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a4977cb8ca54638f22a60675bfbef71bbb77ab6&vid=942501aaa5afc8d85b82628854aac94d517322eb&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay Min. 99.0 %
Loss on ignition (800°C) Max. 0.5 %
Water insoluble matter Max. 0.01 %
pH (5 %, 25 °C) 5.2 - 9.2
Calcium (Ca) Max. 0.01 %
Iron (Fe) Max. 0.001 %
Potassium (K) Max. 0.01 %
Magnesium (Mg) Max. 0.005 %
Heavy metals (as Pb) Max. 5 ppm
Chloride (Cl) Max. 10 ppm
Phosphate (PO₄) Max. 0.001 %
Total N Max. 5 ppm
Grain Size 10 - 60 Mesh Min. 85 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR