Economy water vapor trap
Leverandør: Merck
23988
23988.EA 2720 NOK
23988.
Economy water vapor trap
Filter Gassfilter
This economy water vapor trap is designed for moisture removal for compressed air service to pneumatically-controlled devices.
  • Purifier body made of clear polycarbonate
  • Trap contains a mixture of Molecular Sieves 4A and 13X
  • Not to be used with carrier gases due to permeability of purifier body