Bariumklorid 20% in aqueous solution

Leverandør: VWR Chemicals
Warning

240314.5000EA 1575 NOK
240314.5000
Bariumklorid 20% in aqueous solution
Bariumklorid
Formel: BaCl₂
Kokepunkt: ∼100 °C (1013 hPa)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003445
CAS-nummer: 10361-37-2
EINECS: 233-788-1
FN: 3287
ADR: 6.1,III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR