Jern ≥98%, powder, Technical Grade

Leverandør: VWR Chemicals
Warning

24086.292EA 1700 NOK
24086.292
Jern ≥98%, powder, Technical Grade
Jern
Formel: Fe
Kokepunkt: 3000 °C (1013 hPa)
Smeltepunkt: 1536 °C
Tetthet: 7,86 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00010999
CAS-nummer: 7439-89-6
EINECS: 231-096-4
FN: 3089
ADR: 4.1,III

Bestill nå

Spesifikasjon Testresultater

Assay Min. 98 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR