VWR®, buet basestativ
Leverandør: VWR Collection
241-0104EA 5275 NOK
241-0104
VWR®, buet basestativ
Stativer
PU coated cast iron stand, high grade steel rod 500×12 mm.
  • Special shape of the stand ensures it is particularly solid and steady
The rod can be removed, has M8 thread, and is fixed from below with a washer and a nut.