VWR®, Bossheads, Universal, Pivoting
Leverandør: VWR Collection
241-0113EA 1575 NOK
241-0113
VWR®, Bossheads, Universal, Pivoting
Klemmer Kryssmuffer
Bosshead made of laboral aluminium alloy for connecting two retort rods at any angle.
  • Tommy screw for securing the bosshead at the desired angle
  • T-screws for securing the retort rods