VWR®, Large H Stands (B)
Leverandør: VWR Collection
241-0116EA 5925 NOK
241-0116 241-0117
VWR®, Large H Stands (B)
Stativer
H-shaped stands with nickel-coated steel rod and 420×400 mm grey marbled cast iron baseplate.
  • H-shaped stand has adjustable baseplate
  • Designed to accommodate the upright in any one of three positions