Stativfot, trefot, uten stang
Leverandør: USBECK Laborgeräte
2040 2041
241-0247EA 1450 NOK
241-0247 241-0248
Stativfot, trefot, uten stang
Stativer
Powder-coated steel, thickness 10 mm.
  • For two rods up to 16 mm Ø
  • Hinged screws M6
  • Slip resistant adjusting feet