Separating funnel holder
Leverandør: Brand
140300
241-0345EA 245 NOK
241-0345
Separating funnel holder
Stativer
PP, white.
For conical separating funnels up to 1000 ml. Easely mounted on support rods with an outer Ø 8 to 14 mm with a locking nut.