Clamps, universal, slewable
Leverandør: USBECK Laborgeräte
2508
241-0349EA 670 NOK
241-0349
Clamps, universal, slewable
Klemmer Universalklemmer
Universal clamp made of powder-coated zinc die-cast.With cork lined jaws.
  • Capacity 0 to 80 mm
  • Slewing range 180°