VWR® Talon®, Swivel Clamps
Leverandør: VWR Collection
241-0410EA 610 NOK
241-0410 241-0407 241-0408
VWR® Talon®, Swivel Clamps
Klemmer Universalklemmer
Nickel plated zinc or electro-polished stainless steel. These swivel clamps have an integral holder for attaching to a lab frame or other apparatus. The shaft wing nut allows the holding angle of the swivel clamp to adjust through 360° of rotation.
  • Holds apparatus near the lab frame
  • For circular or irregular objects
  • Built in holder grips rods up to 19 mm in Ø
  • Holder is adjustable for forward or reverse facing adjustment screws
  • Can be locked in place once desired position is achieved
Leveringsinformasjon: Supplied with non-slip vinyl sleeves andMore Product Information