VWR® Talon®, Clamps, Fixed Position, Two Pronged
Leverandør: VWR Collection
241-0409EA 1550 NOK
241-0409
VWR® Talon®, Clamps, Fixed Position, Two Pronged
Klemmer Universalklemmer
Fixed-position clamps made of stainless steel with electro-polished finish. Used to hold apparatus near the lab frame where no adjustment is required after set up.
  • With non-slip vinyl sleeves
  • Built-in holder grips rods up to 19 mm Ø
  • Can be rotated after attachment to a lab frame or other apparatus
Leveringsinformasjon: Supplied with non-slip vinyl sleeves, and with fiberglass covers for temperatures above 100 °C (212 °F).