Capillary column, Omegawax®, Supelco®
Leverandør: Merck
24152
24152.EA 10000 NOK
24152.
Capillary column, Omegawax®, Supelco®
Kromatografikolonner
This column with bonded poly(ethylene glycol) phase allows highly reproducible analyses of fatty acid methyl esters (FAMEs), specifically the omega 3 and omega 6 fatty acids. It is tested to ensure reproducible FAME equivalent chain length (ECL) values and resolution of key components.
Temperature limits: 50 - 280 °C
Sertifiseringer: Meets USP G16 requirements.