Capillary columns, traditional phases, SAC-5, Supelco®
Leverandør: Merck
24156
24156.EA 9160 NOK
24156.
Capillary columns, traditional phases, SAC-5, Supelco®
Kromatografikolonner
This non-polar column is specially designed for reproducible analyses of plant sterols, cholesterol, and other animal sterols.
  • Bonded, Poly(5% diphenyl/95% dimethyl siloxane)
  • High thermal stability - maximum temperatures up to −60 to 320 °C