Jern(II,III)oksid, black, TECHNICAL

Leverandør: VWR Chemicals
Warning

Synonymer: Magnetite, Triiron tetraoxide, Iron(II,III) oxide

24187.460EA 5775 NOK
24187.460
Jern(II,III)oksid, black, TECHNICAL
Jern(II,III)oksid
Formel: Fe₃O₄
MW: 231.54 g/mol
Smeltepunkt: 1560 °C
Tetthet: 5.18 g/cm³ (20 °C)
MDL nummer: MFCD00011010
CAS-nummer: 1317-61-9
EINECS: 215-277-5
Merck Indeks: 12,04087

Bestill nå

Spesifikasjon Testresultater

Identification Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR