Jern(II)sulfat heptahydrat ≥96%, GPR RECTAPUR®

Leverandør: VWR Chemicals
Warning

24240.295EA 610 NOK
24240.295 24240.364
Jern(II)sulfat heptahydrat ≥96%, GPR RECTAPUR®
Jern(II)sulfat heptahydrat
Formel: FeSO₄·7H₂O
Smeltepunkt: 64 °C
Tetthet: 1,9 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149719
CAS-nummer: 7782-63-0
EINECS: 231-753-5
Merck Indeks: 13,04091

Bestill nå

Spesifikasjon Testresultater

Assay Min. 96 %
Not precipitated by NH4OH (as SO4) Max. 0.5 %
Cl (Chloride) Max. 50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR