Florisil® 60-100 mesh for pesticide residue analysis

Leverandør: VWR Chemicals

Synonymer: Magnesium silicate, aktivert, Florisil

24279.185EA 1100 NOK
24279.185
Florisil® 60-100 mesh for pesticide residue analysis
Magnesium silicate
Formel: MgO₃Si
MDL nummer: MFCD00078249
CAS-nummer: 1343-88-0
EINECS: 215-681-1

Bestill nå

Spesifikasjon Testresultater

Identification Passes test

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR