L(+)-Cystein, Millipore®

Leverandør: Merck Millipore Calbiochem
Warning

243005-1
243005-1EA 16875 NOK
243005-1
L(+)-Cystein, Millipore®
L-Cystein
Formel: HSCH₂CH(NH₂)CO₂H
Kokepunkt: 254 °C (1013 hPa)
Smeltepunkt: 115 °C
Tetthet: 1,523 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00064306
CAS-nummer: 52-90-4
EINECS: 200-158-2
Merck Indeks: 13,02810

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR