D(+)-Galactose ≥98.5%, GPR RECTAPUR®

Leverandør: VWR Chemicals

Synonymer: D-Galactose, (2R,3S,4S,5R)-2,3,4,5,6-Pentahydroxyhexanal, (2R,3S,4S,5R)-Pentahydroxyhexanal

24333.296EA 13300 NOK
24333.296
D(+)-Galactose ≥98.5%, GPR RECTAPUR®
D(+)-Galactose
Formel: C₆H₁₂O₆
MW: 180.16 g/mol
Kokepunkt: 527
Smeltepunkt: 163…169 °C
Tetthet: 1.5 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00151230
CAS-nummer: 59-23-4
EINECS: 200-416-4
Merck Indeks: 13,04356
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=508b1d21a6801ddfb1a5f13a64cb06e6f95e8922&vid=3c02003b1d12c9bfd11c2e6faf5cc37d4325d208&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay Min. 98.5 %
Specific optical rotation (10 %; water) 79 - 82 °
Heavy metals (as Pb) Max. 10 ppm
Ignition residue (SO4) Max. 0.1 %
Loss on drying (100°C) Max. 1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR