Magnesiumklorid heksahydrat 99.0-101.0%, krystaller FCC, USP, J.T.Baker®

Leverandør: Avantor

2448-01EA 2375 NOK
2448-01 2448-05 2448-07 2448-R
Magnesiumklorid heksahydrat 99.0-101.0%, krystaller FCC, USP, J.T.Baker®
Magnesiumklorid heksahydrat

  • Food GMP manufactured product

Formel: MgCl₂·6H₂O
Kokepunkt: 1412 °C (1013 hPa)
Smeltepunkt: 117 °C
Tetthet: 1,57 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149781
CAS-nummer: 7791-18-6
EINECS: 232-094-6
Merck Indeks: 13,05684

Bestill nå

Spesifikasjon Testresultater

Meets F.C.C. Requirements 99.0 - 101.0%
Meets U.S.P Requirements Passes Test
Food GMP Manufactured Product Passes Test
GMP Manufactured Product 4.5 - 7.0
Preserve in Tight Containers ≤0.005%
CAUTION: For Manufacturing, processing or repackaging ≤0.005%
Bulk Food Chemical Passes Test
Bulk Pharmaceutical Chemical ≤0.01%
USP - Assay 99.0 - 101.0 %
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - pH (1 in 20) 4.5 - 7.0
USP - Insoluble Matter ≤0.005 %
USP - Sulfate (SO₄) ≤0.005 %
USP - Barium (Ba) Passes Test
USP - Calcium (Ca) ≤0.01 %
USP - Potassium (K) Passes Test
FCC - Assay 99.0 - 101.0 %
FCC - Ammonium (NH₄) ≤0.005 %
FCC - Sulfate (SO₄) ≤0.005 %
FCC - Lead (Pb) ≤4 mg/kg
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR