Aceton 99.5-100.5% (by anhydrous basis) NF, ACS, J.T.Baker®

Leverandør: Avantor
Danger

Synonymer: 2-Propanone, DMK, Propanone, Dimethyl ketone

2462-03EA 0 NOK
2462-03
Aceton 99.5-100.5% (by anhydrous basis) NF, ACS, J.T.Baker®
Aceton
Formel: CH₃COCH₃
MW: 58.08 g/mol
Kokepunkt: 56.2
Smeltepunkt: -95.4
MDL nummer: MFCD00008765
CAS-nummer: 67-64-1
FN: 1090
ADR: 3 II
Merck Indeks: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Meets N.F. Requirements
GMP Manufactured Product
Meets ACS Reagent Chemical Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical Passes Test
NF - Identification A Passes Test
NF - Identification B Passes Test
NF - Specific Gravity at 25°/25°C ≤0.789
NF - Nonvolatile Residue ≤0.004 %
NF - Readily Oxidizable Substances Passes Test
NF - Water (H₂O) ≤0.5 %
NF - Assay (as (CH₃)₂CO) (calculated on anhydrous basis) 99.5 - 100.5 %
NF - Benzene ≤2 ppm
ACS - Aldehydes (as HCHO) ≤0.002 %
ACS - Assay (CH₃COCH₃) (by GC, corrected for water) ≥99.5 %
ACS - Color (APHA) ≤10
ACS - Isopropyl Alcohol (CH₃CHOHCH₃)(by GC) ≤0.05 %
ACS - Methanol (CH₃OH)(by GC) ≤0.05 %
ACS - Residue after Evaporation ≤0.001 %
ACS - Solubility in H₂O Passes Test
ACS - Substances Reducing Permanganate Passes Test
ACS - Titrable Acid (meq/g) ≤0.0003
ACS - Titrable Base (meq/g) ≤0.0006
ACS - Water (H₂O) ≤0.5 %
Class 1, 2 and 3 residual solvents are likely to be present. Benzene (Class 1) is reported on each lot and is below the USP/NF limit concentration. Methanol and Toluene (Class 2) are both below their
Option 1 limits. All Class 3 solvents are <0.5%.
Elemental Impurities (USP (232), EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR