Castor olje, GPR RECTAPUR®

Leverandør: VWR Chemicals

24667.290EA 2075 NOK
24667.290 24667.368
Castor olje, GPR RECTAPUR®
Castor olje
Kokepunkt: 313 °C (1013 hPa)
Smeltepunkt: –10 °C
Tetthet: 0,95 g/cm³ (20 °C)
Flash punkt: >113 °C (closed cup)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00130746
CAS-nummer: 8001-79-4
EINECS: 232-293-8
Merck Indeks: 13,01908

Bestill nå

Spesifikasjon Testresultater

Solubility in methanol Passes test
Acid value Max. 1
Density (20/4) 0.950 - 0.970
Iodine value 80 - 95
Saponification value 170 - 190
Non-saponifiable matter Max. 0.8 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR