Hydraziniumsulfat ≥99.0%, AnalaR NORMAPUR® analytical reagent

Leverandør: VWR Chemicals
Danger

Synonymer: Hydrazine sulphate salt, Hydrazine sulphate

24696.186EA 565 NOK
24696.186
Hydraziniumsulfat ≥99.0%, AnalaR NORMAPUR® analytical reagent
Hydrazinium sulphate
Formel: NH₂NH₂·H₂SO₄
Smeltepunkt: 254 °C
Tetthet: 1,37 g/cm³ (20 °C)
MDL nummer: MFCD00044873
CAS-nummer: 10034-93-2
EINECS: 233-110-4
FN: 3288
ADR: 6.1,III
Merck Indeks: 14,04772
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e277940036b3cf71839ff7458f0c7d08bff6f6d5&vid=7a677e6f1725cfa1b9bdef05cc04304e6739079e&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay Min. 99.0 %
Heavy metals (as Pb) Max. 10 ppm
Ignition residue Max. 0.05 %
Insolubility in water Max. 50 ppm
Cl (Chloride) Max. 50 ppm
Fe (Iron) Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR