Eddiksyre 36,0 - 37,0% NF, Macron Fine Chemicals™

Leverandør: Avantor Performance Materials

Synonymer: Vinegar acid, Methanecarboxylic acid, Ethanoic acid

2488-44EA 2525 NOK
2488-44
Eddiksyre 36,0 - 37,0% NF, Macron Fine Chemicals™
Eddiksyre glasial (iseddik)
Formel: H₃CCOOH
MW: 60.05 g/mol
Kokepunkt: 118
Smeltepunkt: 17
Flash punkt: 38.5
MDL nummer: MFCD00036152
CAS-nummer: 64-19-7
FN: 2790
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

GMP Manufactured Product
Meets N.F. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical 36.0 - 37.0 %
NF - Assay (CH₃COOH) 36.0 - 37.0 %
NF - Chloride (Cl) Passes Test
NF - Identification Passes Test
NF - Nonvolatile Residue ≤0.005 %
NF - Readily Oxidizable Substances Passes Test
NF - Sulfate (SO₄) Passes Test
Appearance (clear, colorless liquid)
Preserve in tight containers, and store at room temperature.
Residual Solvents: Only the Class 3 Solvents acetone, isopropyl acetate,
and propyl acetate are expected to be present. Residual concentration of all Class 3 Solvents combined is <0.1%.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR