L(+)-Glutamin solution 200 mM in water (29.20 mg/ml) cell culture reagent, Corning®

Leverandør: Corning

Synonymer: Gln, L-Glutamine, H-Gln-OH, (S)-(+)-Glutamine, (S)-2,5-Diamino-5-oxopentanoic acid, Levoglutamide, Q

25-005-CI
25-005-CIEA 1350 NOK
25-005-CI
L(+)-Glutamin solution 200 mM in water (29.20 mg/ml) cell culture reagent, Corning®
L(+)-Glutamin

L-glutamine is an essential amino acid and a key component of culture media, serving as a major energy source for propagating cells. It is very stable as a dry powder and as a frozen solution but degrades rapidly in liquid media or stock solutions, producing toxic compounds. Optimal cell performance usually requires supplementation of the media with L-glutamine prior to use. This formulation is prepared in cell culture grade water.


NB: Storage -25°C to -15°C

Formel: C₅H₁₀N₂O₃
MW: 146.15 g/mol
Kokepunkt: 355
MDL nummer: MFCD00008044
CAS-nummer: 56-85-9
Merck Indeks: 13,04484

Bestill nå

Spesifikasjon Testresultater

pH 5.5 ± 0.5
Osmolality 450 ± 50 mOsm/kg H2O
Endotoxin < 0.25 EU/ml @ 1 :50
Mycoplasma Tested Negative
Sterility Pass
Cell Growth Pass

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR