Corning® Trypsin
Leverandør: Corning
25-050-CI
25-050-CIEA 915 NOK
25-050-CI
Corning® Trypsin
Enzymer
0,25% Trypsin in HBSS (Hank’s Balanced Salt Solution) without calcium and magnesium.
  • Porcine parvovirus tested
  • 6×100 ml
Trypsin is an enzymatic reagent that is used to release adherent cells from tissue plates for passaging. This offering contains trypsin at a concentration of 0,25%.