Corning® Trypsin
Leverandør: Corning
25-053-CI
25-053-CIEA 1325 NOK
25-053-CI
Corning® Trypsin
Enzymer
0,05% Trypsin/0,53 mM EDTA in HBSS (Hank’s Balanced Salt Solution) with sodium bicarbonate, without calcium and magnesium.
  • Trypsin EDTA 1X 0,05% Trypsin/0,53 mM EDTA in HBSS with sodium bicarbonate, without calcium and magnesium
  • Porcine Parvovirus tested
Trypsin is used to enzymatically release adherent cells from tissue culture plates for passaging. Divalent cations such as calcium and magnesium, which are often present in the cell culture environment, inhibit this action. EDTA sequesters these ions and thus enhances the efficacy of trypsin. This product contains trypsin at a concentration of 0,05% with 0,53 mM EDTA, and does not contain sodium bicarbonate.
Bestill nå
SPESIFIKASJONER
Enzymnavn Trypsin
Environmentally Preferable
Kilde
Arter
CAS-nr 9002-07-7