Cell dissociation reagent, Corning® Cellstripper®
Leverandør: Corning
25-056-CI
25-056-CIEA 1325 NOK
25-056-CI
Cell dissociation reagent, Corning® Cellstripper®
Cellekultursupplementer og tilsetninger
Cellstripper® is a non-enzymatic cell dissociation solution formulated with a proprietary mixture of chelators that gently dislodge adherent cells in tissue culture. It is an alternative to trypsin when used with either serum-free or serum-supplemented media. Because Cellstripper® is a gentler reagent than trypsin, cells can be exposed to Cellstripper for longer periods of time. Cellstripper® achieves superior results in comparison to enzymatic dissociation products.