Peqlab peqGOLD, DNA Ladders, 50 bp
Leverandør: VWR Chemicals
25-2000EA 1050 NOK
25-2000
Peqlab peqGOLD, DNA Ladders, 50 bp
Elektroforese reagenser Molekylvekt markører
DNA ladder for quantitative and qualitative analysis of dsDNA.
  • Range: 13 distinct fragments 50 to 1000 bp in size
  • Two highlighted bands for easy orientation
  • Also available as ready to use mix containing Orange G dye instead of bromophenol blue