illustra™ Ready-To-Go™ GenomiPhi™ HY DNA amplification kits
Leverandør: Cytiva
25-6603-24 25-6603-96
25-6603-24EA 6150 NOK
25-6603-24 25-6603-96
illustra™ Ready-To-Go™ GenomiPhi™ HY DNA amplification kits
Nukleinsyre reagenser Isothermal Amplification Reagents
illustra™ Ready-To-Go™ GenomiPhi™ HY DNA Amplification Kit offers highly efficient and representative whole-genome amplification with 40 to 60 μg yield from nanogram amounts of DNA sample.
  • Representative amplification of the whole genome
  • Preformulated, predispensed, single dose, lyophilised cakes for rapid reaction set-up and greater reproducibility
  • Quick and simple automation-friendly protocol; no thermal cycler required
  • Shipping and storage at room temperature provides easy access and reduced carbon footprint
  • Yields high quality DNA (eg. for high-throughput genotyping and DNA archival)
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Applikasjon