Dimetylsulfoksid, sterile filtered

Leverandør: Corning
Warning

Synonymer: Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO

25-950-CQC
25-950-CQCEA 1300 NOK
25-950-CQC
Dimetylsulfoksid, sterile filtered
Dimetylsulfoksid

Dimethyl Sulfoxide (DMSO) is commonly used in cell cryopreservation, along with glycerol, to prevent damage to the cell membrane during freezing.


  • Endotoxin tested
  • Septum cap
  • Shelf life 12 months


Mediatech, lnc. products are prepared by a validated aseptic sterile filtration process to ensure that ali products comply with the industry's Sterility Assurance Level (SAL) of at least 1 o-3 (i.e. demonstrating th at a product produced in this manner has no more th an 1 random contaminant per 1000 units).
This product was tested in accordance with currently approved Mediatech, lnc. specifications and procedures. Testing procedures are maintained in compliance with the current versions of the USP and/or EP , where applicable.
For Research Use Only. Not for Human Therapeutic Use. Utilization of this product apart from the labeled intended use may be a violation of local and/or Federal Law.

Formel: (CH₃)₂SO
MW: 78.14 g/mol
Kokepunkt: 189 °C (1013 hPa)
Smeltepunkt: 18.5 °C
Tetthet: 1.101 g/cm³ (20 °C)
Flash punkt: 87 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002089
CAS-nummer: 67-68-5
Merck Indeks: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR