Magnesium ≥99.0%, ribbon, GPR RECTAPUR®

Leverandør: VWR Chemicals
Danger

25071.132EA 1075 NOK
25071.132
Magnesium ≥99.0%, ribbon, GPR RECTAPUR®
Magnesium

Length: approx. 22 m
Width: approx. 3 – 4 mm
Thickness: approx. 0.3 – 0.4 mm

Formel: Mg
Kokepunkt: 1107 °C (1013,25 hPa)
Smeltepunkt: 651 °C
Tetthet: 1,75 g/cm3 (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00085308
CAS-nummer: 7439-95-4
EINECS: 231-104-6
FN: 1869
ADR: 4.1,III
New Product

Bestill nå

Spesifikasjon Testresultater

Assay (Magnesium) min. 99.0%
Iron (Fe) max. 0.01%
Lead (Pb) max. 0.01%
Manganese (Mn) max. 0.030%
Nickel (Ni) max. 0.005%
Zinc (Zn) max. 0.01%

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR